Basic fuchsin 70+%, pure, certified
Product TS40169-1000 is a replacement for TS40169-0250 but varies by Brand, Packaging or Quantity.
Product Details & Documents
- CAS Number:632-99-5
- Formula:C₂₀H₂₀ClN₃
- MDL Number:MFCD00012569
- Molecular Weight:337.85 g/mol
- Merck Index:12,05686
- Melting Pt:250 °C (dec.)
- Flash Pt:200 °C
- Storage Temperature:Ambient
- EINECS:211-189-6
Specifications
Specification Test Results
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥70.0 %
- UV Lambda max:547 to 552 nm (in 50% ethanol)
- Absorption Ratio P-15/P+15:1.16 to 1.35
- BSC Certification:Passes
Specifications
- Pack type:Glass bottle for solids
- Size:25 g
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



