Polysorbate 20
Glass bottle, 250 ml
Catalog # TS23336-2500
Supplier: Thermo Scientific Chemicals
: 23336-2500
Jump to:
Product Details & Documents
Synonyms: Polyoxyethylene-20-Sorbitan Monolaurate, Sorbitan monolaurate, ethoxylated
- CAS Number:9005-64-5
- Formula:C₅₈H₁₁₄O₂₆
- MDL Number:MFCD00165986
- Boiling Pt:>100 °C (760 mmHg)
- Density:1.1 g/cm³
- Flash Pt:>150 °C
- Storage Temperature:Ambient
- EINECS:500-018-3
Specifications
Specification Test Results
- Appearance:Clear to slightly hazy yellow to brown-yellow Viscous liquid (May develop some turbidity or precipitate)
- Infrared spectrum:Conforms
- Refractive index:1.4680 to 1.4720 (20°C, 589 nm)
- Water:≤3 %
- Dioxane:≤10 ppm
- Acid value:≤2 mg KOH/g
- Hydroxyl value:96 to 108 mg KOH/g
- Saponification value:40 to 50 mg KOH/g
Specifications
- Pack type:Glass bottle
- Size:250 ml
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



