Polydimethylsiloxane, 100 cSt
Glass bottle, 2.5 kg
Catalog # TS17844-0025
Supplier: Thermo Scientific Chemicals
Supplier #: 17844-0025
Polydimethylsiloxane, 100 cSt
Catalog # TS17844-0025
Supplier: Thermo Scientific Chemicals
CAS Number: 63148-62-9
Jump to:
Product Details & Documents
Synonyms: Dimethylpolysiloxane, PDMS, Silicone oil, Dimethicone
- CAS Number:63148-62-9
- Formula:(CH₃)₃SiO(Si(CH₃)₂O)n
- MDL Number:MFCD00084411
- Boiling Pt:>65 °C (760 mmHg)
- Density:0.965 g/cm³
- Flash Pt:>100 °C
- Storage Temperature:Ambient
Documents
Certificates
Search by Lot NumberSpecifications
Specification Test Results
- Appearance:Clear viscous liquid
- Infrared spectrum:Conforms
- Refractive index:1.4020 to 1.4050 (20°C, 589 nm)
- Color scale:≤35 APHA
- Viscosity:95 to 105 cSt (at 25°C)
- Volatiles:≤0.5 % (200°C)
Specifications
- Pack type:Glass bottle
- Size:2.5 kg
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



