Diethyl ether 99+% stabilised, extra pure
Metal drum, 5 L
Catalog # TS17683-0050
Supplier: Thermo Scientific Chemicals
: 17683-0050
Diethyl ether 99+% stabilised, extra pure
Catalog # TS17683-0050
Supplier: Thermo Scientific Chemicals
CAS Number: 60-29-7
Jump to:
Product Details & Documents
Synonyms: Ethyl Ether, Ether, 2,2'-Oxydiethane, 2,2'-Oxybisethane, Ethoxyethane
- CAS Number:60-29-7
- Formula:(CH₃CH₂)₂O
- MDL Number:MFCD00011646
- Molecular Weight:74.12 g/mol
- Boiling Pt:34.6 °C (760 mmHg)
- Melting Pt:–116 °C
- Density:0.714 g/cm³
- Flash Pt:–45 °C
- Storage Temperature:Ambient
- UN:1155
- ADR:3,I
- :Stabilised with BHT (2,6-Di-Tert-Butyl-4-Methylphenol; Ionol)
Specifications
Specification Test Results
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.3510 to 1.3540 (20°C, 589 nm)
- Water:≤0.1 % (Coulometric)
- GC:≥99.0 %
- Color scale:≤10 APHA
- Stabilizer:5 to 7 ppm BHT
- Relative density:(20°C) 0.714 to 0.716
- Residue after evaporation:≤0.005 %
- Acidity:≤0.001 meq/g
- Peroxides:≤0.0005 %
Specifications
- Pack type:Metal drum
- Size:5 L
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



