Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Acrylamide 98.5%, extra pure

Acrylamide 98.5%, extra pure

Plastic bottle, 2.5 kg
Catalog # TS16483-0025
Toxic
Health
Supplier #:  16483-0025
Acrylamide 98.5%, extra pure
Catalog # TS16483-0025
CAS Number:  79-06-1
Acrylamide 98.5%, extra pure
Acrylamide 98.5%, extra pure

Plastic bottle, 2.5 kg

CAS Number:  79-06-1
Price
$326.00
Log in for availability
Each (2,500g)
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
Synonyms:  Prop-2-enamide
  • CAS Number:79-06-1
  • Formula:C₃H₅NO
  • MDL Number:MFCD00008032
  • Molecular Weight:71.08 g/mol
  • Boiling Pt:125 °C (25 mmHg)
  • Melting Pt:82…86 °C
  • Density:1.122 g/cm³
  • Flash Pt:138 °C
  • Storage Temperature:Refrigerator
  • EINECS:201-173-7
  • UN:2074
  • ADR:6.1,III
Specifications

Specification Test Results

  • Appearance:
    White Crystalline powder or crystals
  • Infrared spectrum:
    Conforms
  • Melting point:
    82 to 86 °C
  • Water:
    ≤2.0 %
  • GC:
    ≥98.0 %
  • Color of solution:
    (10 % in water) Clear and colorless

Specifications

  • Pack type:
    Plastic bottle
  • Size:
    2.5 kg
  • Cat. No.:
    TS16483-0025

Specifications

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.