2-Phenoxyethanol 99%
25 L, Metal drum
Catalog # TS13024-0250
Supplier: Thermo Scientific Chemicals
Supplier #: 13024-0250
2-Phenoxyethanol 99%
Catalog # TS13024-0250
Supplier: Thermo Scientific Chemicals
CAS Number: 122-99-6
Jump to:
Product Details & Documents
Synonyms: ω-Hydroxyphenetole, β-Hydroxyphenetole, Phenyl cellosolve, Phenoxyethanol, Ethylene glycol monophenyl ether
- CAS Number:122-99-6
- Formula:C₆H₅OCH₂CH₂OH
- MDL Number:MFCD00002857
- Molecular Weight:138.17 g/mol
- Merck Index:12,07410
- Boiling Pt:237 °C (760 mmHg)
- Melting Pt:11…13 °C
- Density:1.1 g/cm³
- Flash Pt:130 °C
- Storage Temperature:Ambient
- EINECS:204-589-7
Specifications
Specification Test Results
- Appearance:Clear colorless Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5360 to 1.5400 (20°C, 589 nm)
- GC:≥98.5 %
- pH:9.5 to 11.0 (5% at 25°C)
- Titration with HClO4:≥99 %
- Iron (Fe):≤0.001 %
Specifications
- Pack type:Metal drum
- Size:25 L
Specifications
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.



