Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
VWR® Wire Frame Shelves

VWR® Wire Frame Shelves

VWR® full wire frame shelf for 14 cu. ft. units, 76498-992, 76499-006, 76499-000, 76177-834, 76177-832, 76498-986, Wire, 483×406×51 mm (19×16×2"), 2.3 kg (5 lbs.)
Catalog # 76539-092
Supplier:   VWR International
VWR® Wire Frame Shelves
Catalog # 76539-092
Supplier:  VWR International
VWR® Wire Frame Shelves
VWR® Wire Frame Shelves

VWR® full wire frame shelf for 14 cu. ft. units, 76498-992, 76499-006, 76499-000, 76177-834, 76177-832, 76498-986, Wire, 483×406×51 mm (19×16×2"), 2.3 kg (5 lbs.)

Price
$310.77
Log in for availability
Each
Product Details & Documents

Extra wire frame shelves that fit the VWR® 14, 17, and 20 cu. ft. refrigerators.

Specifications
  • Description:
    VWR® full wire frame shelf for 14 cu. ft. units
  • Material:
    Wire
  • Exterior W×D×H:
    483×406×51 mm (19×16×2")
  • For use with:
    76498-992, 76499-006, 76499-000, 76177-834, 76177-832, 76498-986
  • Weight:
    2.3 kg (5 lbs.)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.