Order Entry
Puerto Rico
ContactUsLinkComponent
Masterflex® Dispensing Gun PTFE 3/8"npt (Spray Gun Valves (actuated))
Masterflex® Dispensing Gun PTFE 3/8"npt (Spray Gun Valves (actuated))
Catalog # MFLX98515-05
Supplier:  DYTEC CORPORATION
undefined
Masterflex® Dispensing Gun PTFE 3/8"npt (Spray Gun Valves (actuated))
Catalog # MFLX98515-05
Supplier:  DYTEC CORPORATION
Supplier Number:  THG-LD6P-TFEV

About this item

No splashing.

  • Recirculating gun has a contamination free fluid path to eliminate bacterial growth
  • Highly resistant to corrosive substances and fumes
  • Spray guns have a 1 GPM flow rate; max operating pressure of 75 psi

These Masterflex® Tank Dispensing Spray Guns feature a special tip to eliminate splashing. Choose from PTFE or stainless steel construction for a wide range of applications.

  • Description:
    Dispensing gun PTFE 3/8"npt (spray gun valves (actuated))
  • Environmentally Preferable:
  • Wetted Materials:
    PTFE
  • Type:
    98515-05
  • Warranty:
    1 year
  • Connection Type:
    NPT(F)
  • Cat. No.:
    MFLX98515-05