Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Easy-Load® 3 Pump Head Mounting Hardware, Mounts One Pump Head, Stainless steel, Masterflex L/S® Easy-Load® 3 Pump head, L/S
Catalog # MFLX77800-00
Supplier: Avantor Fluid Handling
Supplier #: 77800-00
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Catalog # MFLX77800-00
Supplier: Avantor Fluid Handling
Jump to:
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
- Constructed of durable stainless steel
- See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Specifications
- Description:L/S® Easy-Load® 3 Pump Head Mounting Hardware, Mounts One Pump Head
- Material:Stainless steel
- Compatible with:Masterflex L/S® Easy-Load® 3 Pump head
- Pump Series:L/S