Orders LinkContactUsLinkComponent
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Easy-Load® II Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty, Stainless steel, Masterflex L/S® Easy-Load® II Pump head, L/S
Catalog # MFLX77200-01
Supplier #:  77200-01
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Catalog # MFLX77200-01
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
Masterflex® L/S® Pump Head Mounting Hardware, Avantor®
L/S® Easy-Load® II Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty, Stainless steel, Masterflex L/S® Easy-Load® II Pump head, L/S
Price
$59.00
Log in for availability
Each
Product Details & Documents
Easy stack multiple pump heads on your Masterflex® L/S® drive.
  • Constructed of durable stainless steel
  • See drive specifications for recommended number of heads
Stack multiple pump heads or replacement single pump head mounting hardware. Choose mounting hardware based on the Masterflex® L/S® pump head that you have. Compatible with the Masterflex L/S series pumps.
Specifications
  • Description:
    L/S® Easy-Load® II Pump Head Mounting Hardware, One Pump Head, Stainless Steel; 2/Set. 2 years warranty
  • Material:
    Stainless steel
  • Compatible with:
    Masterflex L/S® Easy-Load® II Pump head
  • Pump Series:
    L/S