Orders LinkContactUsLinkComponent
Masterflex® Single-Use Forceps, Avantor®
Masterflex® Single-Use Forceps, Avantor®
Sterile Forceps, stainless steel, 4,5" long; 25/pk, Stainless steel, 114 mm (4.5"), +
Catalog # MFLX75955-13
Masterflex® Single-Use Forceps, Avantor®
Catalog # MFLX75955-13
Supplier:  
Masterflex® Single-Use Forceps, Avantor®
Sterile Forceps, stainless steel, 4,5" long; 25/pk, Stainless steel, 114 mm (4.5"), +
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
DRESSING FORCEP 8IN STER CS25 Catalog #76669-824
Product 76669-824 is a functional alternative to MFLX75955-13 but Packaging, Quantity, Manufacturer and/or specifications may vary.
Product Details & Documents
No more cross-contamination.
  • Protect yourself and your samples with disposable forceps
  • Autoclavable plastic or stainless steel construction
These single-use forceps are bulk-packed for convenience and affordability. Slide forceps feature an extra-wide grip for greater stability and security when staining cells and tissue. Fine-tipped forceps are ideally suited for work with smaller specimens. Stainless steel forceps have grooved finger grips and ridged tips, best for larger, soft or wet specimens.
Specifications
  • Description:
    Sterile Forceps, stainless steel, 4,5" long; 25/pk
  • Material:
    Stainless steel
  • Overall length:
    114 mm (4.5")
  • Sterile:
    +
Accessories
Product Information
  • Catalog #
    Material
    Sterile
    Availability
    Price
  • Adhesive coated microscope slides, Poly-l-lysine
    Adhesive coated microscope slides, Poly-l-lysine
    MFLX75955-45
    Glass
    -
    Log in for availability
    Each - $177.00