Pulsafeeder Tube Assembly Kit for the peristaltic pump 74130-62 and 74130-63 pumps
Catalog # MFLX74130-43
Supplier: EQUIP SOLUTIONS
ETL listed
ISO9001
About this item
Enhance your pumps performance.
- Easily change out tube for optimum performance
- Easy and quick maintenance
Optimize performance of your Pulsafeeder Economical Peristaltic Pump.
- Maximum Flow Rate:43.3 ml/min
- Max. Pressure:6.9 bar (100 psi)
- Environmentally Preferable:
- Warranty:2 years
- Cat. No.:MFLX74130-43