Masterflex® Drum Pump Replacement Motor, ODP; 110V/60Hz
Catalog # MFLX70604-03
Supplier: VWR International
ETL Canada listed
About this item
Safety transfer a variety of liquids from drums, tanks, and small storage vessels.
- Use with Masterflex® Complete Drum and Container Pumps
Drum and container pumps are engineered to safely transfer a variety of liquids from drums, tanks and small storage vessels. These Masterflex® drum pump replacement motors are one component of a system that includes a pump tube and motor (order pump tubes separately). Motors are equipped with variable-speed control for added safety and control.
- Power:110 VAC; 60 Hz
- Description:Replacement motor, ODP, 110 VAC, 60 Hz
- Environmentally Preferable:
- Warranty:3 years
- Motor type:Open drip proof (ODP)
- Cat. No.:MFLX70604-03