Masterflex® luer fitting, male luer lock×1/8 hose barb, Orange, 25/pk"
Catalog # MFLX45520-53
Supplier: VWR International
About this item
Quick and easy assembly without tools.
- Constructed of durable Polyolefin
Available in 6 colors for quick and easy identification: berry, rose, yellow, lime, violet, and orange. Made of polyolefin, a transparent polycarbonate alternative containing no bisphenol-A (BPA).
Superior chemical resistance compared to polycarbonate. Sterilize by autoclave, gamma irradiation, or ethylene oxide. Meets USP Class VI. Animal-derived ingredient-free. Packs of 25.
- Barb size/tubing int.Ø (port 1):⅛"
- Connection Type:Male Luer to Barb
- Flow pattern:Straight
- Description:luer fitting, male luer lock×1/8 hose barb, Orange, 25/pk"
- Environmentally Preferable:
- Connection type (port 1):Male luer lock
- No. of ports:2
- Warranty:None
- Cat. No.:MFLX45520-53
- Fitting Material:CrystalVu