Masterflex® Fitting, Polypropylene, Straight, Female Luer to Thread Adapter, with 5/16" Hex to 1/4-28 UNF; 25/PK
MFLX45508-66
CE certificate
Easily transition between luer fittings and UNF or NPT threads.
- Repackaged in an ISO 9001 facility in an ISO Class 7 Cleanroom
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
- Connection Type: Female Luer to Threaded, with 5/16" Hex to 1/4-28 UNF
These adapters transition between luer fittings and UNF or NPT threads. Both male and female luer connections are available. All threaded connections are female thread.
- Connection Type:Female Luer to Thread
- Flow pattern:Straight
- Description:Fitting, Polypropylene, Straight, Female Luer to Thread Adapter, with 5/16" Hex to 1/4-28 UNF; 25/PK
- Environmentally Preferable:
- Model:45508-66
- Connection type (port 1):Female luer
- Connection type (port 2):1/4-28 UNF thread
- No. of ports:2
- Cat. No.:MFLX45508-66
- Fitting Material:PP
- Thread Size UNF(M) (Port 1):¼"-28