Female Threaded Tee Coupling, 1/4" NPT(F), PTFE
Catalog # MFLX34030-20
Supplier: MICROMOLD PRODUCTS, INC.
About this item
Maintain a permanent connection.
- Ideal for general-purpose and single-use applications, to connect tubing with rest of system
- Determine which connector type is needed for your application by contacting free technical application assistance*
- Global market leader in range of materials for most extensive chemical compatibility
- Single-use custom configuration solutions to minimize downtime
- Regulatory approvals to match your unique application
- Unique packaging options include individuals, packs, and kits for customer-friendly ordering and delivery
Threaded fittings are designed for more permanent installations. Choose the material and size that meets your application needs.
- Connection Type:Threaded Union
- Flow pattern:T
- Thread size NPT(F) (port 1):¼"
- Thread size NPT(F) (port 2):¼"
- Description:Female Threaded Tee Coupling, ¼" NPT(F), PTFE
- Environmentally Preferable:
- Model:34030-20
- Sterilization:Autoclave or ethylene oxide (ETO)
- Cat. No.:MFLX34030-20
- Fitting Material:PTFE
- Thread Size NPT(F) (Port 3):¼"