Order Entry
Puerto Rico
ContactUsLinkComponent
 
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.125" Diameter
  MFLX17315-22
undefined
CINC Centrifugal Multi-Purpose Separator/Extractor Weir; 1.125" Diameter
  MFLX17315-22
 :  02W1.125B-2

 

Mix, react, and/or separate liquid-liquid mixtures in a single device.

  • Highly flexible design operates effectively under changing inlet concentrations, variable flow rates, and flow disruptions
  • Interchangeable weirs allow for broad applications in research, pilot plant, or production operations

Weirs are available in various diameters. Use the large diameter weirs in applications with high-viscosity liquids.

  • Ø:
    2.8575 cm
  • Description:
    Centrifugal Multi-Purpose Separator/Extractor Weir; 1.125" Diameter
  • Environmentally Preferable:
  • Warranty:
    2 years
  • Cat. No.:
    MFLX17315-22