ContactUsLinkComponent
Masterflex® L/S® Gear Service Kits for 6 to 600 rpm Drives
Masterflex® L/S® Gear Service Kits for 6 to 600 rpm Drives
Catalog # MFLX07553-06
undefined
Masterflex® L/S® Gear Service Kits for 6 to 600 rpm Drives
Catalog # MFLX07553-06
Supplier Number:  7553-06
About this item
Make the most of your Masterflex drives replacement parts and service kits.
  • Easy to install, these service kits avoid costly downtime of your gear pump head
  • Suitable for use with Masterflex® L/S® 6 to 600 rpm drives
Extend the life of your Masterflex pump with these replacement parts.
Delivery: Includes gear assembly (nylon gear/shaft), gear case cover gasket, and replacement fuse.
  • Description:
    Service kits for L/S 6-600 rpm drives.
  • Environmentally Preferable:
  • Warranty:
    1 year
  • Compatible with:
    07520-00, -20, -25, -40, -45, 07521-00, -20, -25, -40, -45, 07522-10, -15, 07523-00, -02, -20, -25, 07525-00, -02, -34, -36, 07526-00, -05, 07527-00
  • Cat. No.:
    MFLX07553-06
Frequently Bought Together