Fitting, PFA, Elbow, Threaded Union, 1/2" NPT(F)
Catalog # MFLX06546-35
Supplier: ANDON SPECIALTIES INC.
About this item
Chemically-inert PFA handles most aggressive chemicals.
- Quick and easy assembly without tools
Threaded fittings offer a more permanent installation. Choose the size to meet your application needs.
- Connection Type:Threaded Union
- Flow pattern:Elbow
- Thread size NPT(F) (port 1):½"
- Thread size NPT(F) (port 2):½"
- Description:Fitting, PFA, Elbow, Threaded Union, 1/2" NPT(F)
- Environmentally Preferable:
- Sterilization:Ethylene oxide (ETO) only
- Warranty:1 year
- Cat. No.:MFLX06546-35
- Fitting Material:PFA