Straight threaded coupling, PFA, 1/2" NPT(F)
Catalog # MFLX06546-03
Supplier: ANDON SPECIALTIES INC.
About this item
Create a secure pipe threaded connection.
- Constructed of chemically inert PFA
These straight threaded coupling fittings are made of chemically inert PFA. Fittings can be sterilized via autoclave or ethylene oxide (ETO).
- Connection Type:Threaded Union
- Flow pattern:Straight
- Thread size NPT(F) (port 1):½"
- Thread size NPT(F) (port 2):½"
- Description:Straight threaded coupling, PFA, ½" NPT(F)
- Environmentally Preferable:
- Sterilization:Ethylene oxide (ETO) only
- Warranty:1 year
- Cat. No.:MFLX06546-03
- Fitting Material:PFA