Orders LinkContactUsLinkComponent
SepMate™-50 (IVD)
SepMate™-50 (IVD)
SepMate™-50 (IVD), 500 units
Catalog # MSPP-85460
Marketsource
Supplier #:  85460
SepMate™-50 (IVD)
Catalog # MSPP-85460
Marketsource
SepMate™-50 (IVD)
SepMate™-50 (IVD)
SepMate™-50 (IVD), 500 units
Price
$2,955.38
Log in for availability
Pack of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
50 ml centrifuge tube with an insert for density gradient centrifugation
  • Eliminates the need for carefully layering blood over the density gradient medium (e.g. Lymphoprep™, and more)
  • Reduces total centrifuge time to 10 minutes with the brake on for fresh samples
  • Allows fast and easy harvesting of the isolated mononuclear cells by simply pouring off the supernatant
  • Can be combined with RosetteSep™ enrichment cocktails to isolate specific cell types in just 30 minutes
Simplify peripheral blood mononuclear cell to isolate specific immune cell subsets.
SepMate™ is manufactured under cGMP and registered as an In Vitro Diagnostic on SepMate™.
Packaging: Packed with 100 tubes, dispenser box containing 4 bags, 25 tubes/bag.
Caution: Product restrictions: (i) Customer shall not make products or any portion of them in any way, shape or form, including as a component of another product available for the purpose of further resale or alter or remove the Product label and the Supplier’s mark of origin without express written permission of Supplier and (ii) Customer is familiar with Supplier’s restrictions on the use of the products and shall use the products in compliance with such restrictions and the applicable Supplier Product Information sheet (collectively, the 'Product Restrictions').
Documents
Specifications
  • Capacity:
    500 units
  • Description:
    SepMate™-50 (IVD)