Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco

Vikan® Hygienic Wall Bracket, Double Hook Module, Remco

Pink, 3.27×3.07×1.89"
Catalog # 76528-672
Supplier:   Remco Products
Supplier #:  10141
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco
Catalog # 76528-672
Supplier:  Remco Products
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco
Vikan® Hygienic Wall Bracket, Double Hook Module, Remco

Pink, 3.27×3.07×1.89"

Price
$18.24
Log in for availability
Each
Product Details & Documents

Double hook module is designed for 1-2 cleaning tools that have a hanging hole with a D- or T- shapped handle.

  • Can hold products weighing up to 6.61 lbs.
  • Polypropylene/polyamide construction

The double hook module is easy to disassemble for cleaning or replacement.

Specifications
  • Color:
    Pink
  • Dimensions:
    3.27×3.07×1.89"