Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco

Vikan® Hygienic Wall Bracket, Single Hook Module, Remco

Grey, 1.63×3.07×1.89"
Catalog # 76528-644
Supplier:   Remco Products
Supplier #:  101288
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Catalog # 76528-644
Supplier:  Remco Products
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco

Grey, 1.63×3.07×1.89"

Price
$13.71
Log in for availability
Each
Product Details & Documents

Single hook module is designed for storing cleaning tools that have a hanging hole.

  • Can hold products weighing up to 6.61 lbs.
  • Polypropylene/polyamide construction

The hook system is easy to disassemble for cleaning or replacement.

Specifications
  • Color:
    Grey
  • Dimensions:
    1.63×3.07×1.89"