Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Lime green, 1.63×3.07×1.89"
Catalog # 76528-640
Supplier: Remco Products
Supplier #: 101277
Vikan® Hygienic Wall Bracket, Single Hook Module, Remco
Catalog # 76528-640
Supplier: Remco Products
Jump to:
Product Details & Documents
Single hook module is designed for storing cleaning tools that have a hanging hole.
- Can hold products weighing up to 6.61 lbs.
- Polypropylene/polyamide construction
The hook system is easy to disassemble for cleaning or replacement.
Specifications
- Color:Lime green
- Dimensions:1.63×3.07×1.89"
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



