Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
STORAGE BIN SMALL STACKABLE
Catalog # 76519-978
Supplier:
Luxor
Expand image
STORAGE BIN SMALL STACKABLE
Catalog # 76519-978
Supplier:
Luxor
Supplier Number:
MBS-BIN-8S