Extended Light Path Capillaries, Agilent Technologies
CE extended light path (3x) capillary, bare fused silica, 50 µm/56 cm, bubble factor 3, 150 µm optical path length, Red
AGG1600-61232
: G1600-61232
Extended Light Path Capillaries, Agilent Technologies
AGG1600-61232
Extended light path capillaries that improve sensitivity 3- to 5-fold over standard capillaries.
- Use narrow 25 and 50 µm Int.Ø 'bubble' cell capillaries for highly conductive buffers without sacrificing sensitivity
With extended light path capillaries, the inner diameter is increased only at the detection window, offering the sensitivity of a wide inner diameter capillary and the low current generation of a narrow one. Resolution is not sacrificed when used with matching Agilent optical alignment interfaces.
- Color:Red
- Description:CE extended light path (3x) capillary, bare fused silica, 50 µm/56 cm, bubble factor 3, 150 µm optical path length