Orders LinkContactUsLinkComponent
CrossLab Sample Preparation, Agilent Technologies
CrossLab Sample Preparation, Agilent Technologies
C18 Endcapped Box, 100×1 ml Tubes, 100 mg SampliQ Solid Phase Extraction (SPE)
Catalog # AG5982-1311
Supplier #:  5982-1311
CrossLab Sample Preparation, Agilent Technologies
Catalog # AG5982-1311
CrossLab Sample Preparation, Agilent Technologies
CrossLab Sample Preparation, Agilent Technologies
C18 Endcapped Box, 100×1 ml Tubes, 100 mg SampliQ Solid Phase Extraction (SPE)
Price
$301.00
Log in for availability
Pack of 100
Product Details & Documents
Agilent offers a range of Sample Preparation products to support your workflows.
  • Achieve better results from your analyses, while also extending the lifetime of your column and instrument components
Our suite of sample preparation products can help you to achieve better results from your analyses, while also extending the lifetime of your column and instrument components. Our purification and filtration products provide reliable results for repeated analyses. Our QuEChERS, solid phase extraction (SPE), solid phase microextraction (SPME), and supported liquid extraction (SLE) products ensure your samples are free of interferences, even with complex matrices.
Specifications
  • Description:
    C18 Endcapped Box, 100×1 ml Tubes, 100 mg SampliQ Solid Phase Extraction (SPE)