Orders LinkContactUsLinkComponent
GenScript® GenBox Mini Electrophoresis Tank, GenScript
GenScript® GenBox Mini Electrophoresis Tank, GenScript
GenBox mini blot system
Catalog # 76510-506
Supplier:   Genscript
Supplier #:  L00782
GenScript® GenBox Mini Electrophoresis Tank, GenScript
Catalog # 76510-506
Supplier:  Genscript
GenScript® GenBox Mini Electrophoresis Tank, GenScript
GenScript® GenBox Mini Electrophoresis Tank, GenScript
GenBox mini blot system
Price
$914.23
Log in for availability
Each
Minimum order quantity is 1
  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents
GenBox Gel electrophoresis system is designed to run 1 or 2 mini gels at the same time. GenBox mini tank is designed for ultimate convenience and consistent performance compared to traditional electrophoresis tanks. Discover the versatile and easy to use GenBox mini tank for your gel electrophoresis and blotting needs.
  • Robust: Achieve reproducible protein separation with GenBox
  • Convenience: Run up to 2 mini gels
  • Simple: Easy to set up and operate
  • Efficient: Compatible with Genbox mini blot module for traditional wet transfer
The GenBox mini electrophoresis tank runs both GenScript's 10*8 and 10*10 precast gels interchangeably. It can run one or two mini gels. The mini tank is compatible with GenBox blot modules for tank blotting. The GenBox Mini Trans-Blot module accommodates two gel holder cassettes, which is useful for blotting 2 mini PAGE gels at the same time.
Specifications
  • Description:
    GenBox mini blot system
Accessories
Product Information
  • Catalog #
    Availability
    Price
  • Cassette opener
    Cassette opener
    76198-958
    Log in for availability
    Each - $13.57
Product Information
  • Catalog #
    Availability
    Price
  • Tank adaptor
    Tank adaptor
    76198-956
    Log in for availability
    Each - $14.44