SymLine® Continuous Level Control
Floater, electronic, 139 mm
Catalog # 76503-654
Supplier: SCAT AMERICAS, INC.
Supplier #: 160123
Jump to:
Product Details & Documents
Measuring filling levels continuously. Monitoring non-transparent canisters on a permanent basis is no problem with the continuous level control unit.
- Automatic warning for 5 different filling levels freely adjustable, with accuracy of up to ±0,5 mm
- Control box provides 5 freely configurable outlets to control external devices
- Disc sensor for level control from non-conductive material is easy to install and adjustable in height
- Rod sensor and canister with removable sleeve allow easy maintenance and cleaning
- Canister made from electrostatic conductive HDPE material
The compact design and independent power supply allow simple handling and installation, also outside switch cabinets.
Specifications
- Description:Floater, electronic, 139 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Built-in Signalbox2; 2-channel (EU), for SafetyWasteCap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clampBuilt-in Signalbox2; 2-channel (EU), for SafetyWasteCap and other electronic level controlers, touchless alarm-resetting fields for 2 containers, 2 switch inputs and 2 switch outputs for peripheral units incl. 230 V power supply cable with open end and clamp76502-952Log in for availabilityEach - $1,017.70
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



