SymLine® Grounding and Adapters
Grounding cable with 1 mc connector, 1 connection ring (Ø 5 mm), 1.5 m
Catalog # 76503-570
Supplier: SCAT AMERICAS, INC.
Supplier #: 108272
Jump to:
Product Details & Documents
Dovetail the SymLine® system to suit specific requirements. This product selection includes several useful ancillaries.
- SymLine® offers a suitable adapter for every common thread, to eliminate problems connecting different thread types
- Many useful accessories allow users to configure the SymLine® system according to specific needs
- All accessories are eclectically dissipative and approved for use in laboratories with explosion hazards
Dispose of liquid HPLC waste safely. SymLine® is quickly installed without drilling holes or making technical changes to existing laboratory equipment.
Specifications
- Description:Grounding cable with 1 mc connector, 1 connection ring (Ø 5 mm)
- Length:1.5 m
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



