Orders LinkContactUsLinkComponent
Evergreen Tissue Processing/Embedding Cassettes, Caplugs Evergreen
Evergreen Tissue Processing/Embedding Cassettes, Caplugs Evergreen
4-Compartment biopsy cassette (1 mm holes), Acetal, Grey
Catalog # 76502-848
Supplier:   Caplugs
Supplier #:  258-4249-H90
Evergreen Tissue Processing/Embedding Cassettes, Caplugs Evergreen
Catalog # 76502-848
Supplier:  Caplugs
Evergreen Tissue Processing/Embedding Cassettes, Caplugs Evergreen
Evergreen Tissue Processing/Embedding Cassettes, Caplugs Evergreen
4-Compartment biopsy cassette (1 mm holes), Acetal, Grey
Price
$141.13
Log in for availability
Bag of 500
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Caplugs Evergreen tissue processing/embedding cassettes are used in the processing, embedding, sectioning, and long term storage of tissue and biopsy specimens.
  • High density acetal construction
  • Acetal is resistant to histological solvents such as alcohol and xylene
  • Acetal will also keep the specimen submerged
  • Modified ball-and-socket hinge design for ease of opening and for detachment
  • Cassettes are available unassembled and bulk packed
Tissue processing/embedding cassettes feature efficient flow-through holes (1×1 mm) to maximize fluid exchange and ensure proper drainage.
Specifications
  • Color:
    Grey
  • Description:
    4-Compartment biopsy cassette (1 mm holes)
  • Material:
    Acetal