SymLine® Grounding and Adapters
Seal ring for pipe system
Catalog # 76502-872
Supplier: SCAT AMERICAS, INC.
Supplier #: 106408
Jump to:
Product Details & Documents
Dovetail the SymLine® system to suit specific requirements. This product selection includes several useful ancillaries.
- SymLine® offers a suitable adapter for every common thread, to eliminate problems connecting different thread types
- Many useful accessories allow users to configure the SymLine® system according to specific needs
- All accessories are eclectically dissipative and approved for use in laboratories with explosion hazards
Dispose of liquid HPLC waste safely. SymLine® is quickly installed without drilling holes or making technical changes to existing laboratory equipment.
Specifications
- Description:Seal ring for pipe system