KIT EASYSEP MOUSE PAN-B CELL ISOLATION
Catalog # MSPP-19844A
Supplier: Stemcell Technologies
KIT EASYSEP MOUSE PAN-B CELL ISOLATION
Catalog # MSPP-19844A
Supplier: Stemcell Technologies
Supplier Number: 19844
Frequently Bought Together
TruCool™ Hinged Lid CryoBoxes
Supplier: AZENTA US, INC
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
QUICK-LOAD® Purple DNA Ladder, New England BioLabs
Supplier: New England Biolabs (NEB)
Anti-IgG2a Rat Antibody (FITC (Fluorescein Isothiocyanate)) [clone: SB84a]
Supplier: Southern Biotechnology
KIT AUTOMATIC SETUP BEAD 3UM 2.5X10E6/ML
Supplier: SONY BIOTECHNOLOGY INC. MS
OLIGONUCLEOTIDE 1826 CLASS B CPG TLR9
Supplier: Invivogen
Eppendorf LoBind® twin.tec® PCR Plates
Supplier: Eppendorf
PEANUT AGGLUTININ CY3 RED GALACTOSE 1MG
Supplier: Vector Laboratories
RECOM LFA-3 FC CHIMERA CF >95% HU 50UG
Supplier: R&D Systems
Lympholyte® Mammal Cell Separation Media, Cedar Lane Laboratories
Supplier: Cedar Lane
Ethanol, anhydrous ≥99.9% (200 Proof)
Supplier: Electron Microscopy Sciences
Anti-IgM Rat Monoclonal Antibody (Biotin) [clone: RMM-1]
Supplier: Biolegend