PathOdorizer, Odor Eliminating Solution, Mortech®
Pathodorizer (Deodorizer), 1 gallon
Catalog # 76498-458
Supplier: Mopec
Supplier #: BE040
Jump to:
Product Details & Documents
Developed specially for use by medical examiners, PathOdorizer eliminates offensive odors emitted from decomposed bodies within seconds. From the discovery scene to the morgue, PathOdorizer can be used in every phase of work.
- No CFCs, formaldehyde, or hazardous biocides
- No toxic fumes
- Safe for use on metals, plastics, or fabrics
- PathOdorizer Plus offers additional cleaning and degreasing properties
PathOdorizer not only removes odors from clothes, vehicles, body bags, refrigerators, and other items, it keeps the odor from permeating the lab environment. The formula is completely water soluble, odorless, colorless, biodegradable, non-flammable, and non-corrosive.
Specifications
- Description:Pathodorizer (Deodorizer)
- Size:1 gallon