Orders LinkContactUsLinkComponent
ZawBones Kit and Replacement Components, Mortech®
ZawBones fine replacement blade
  76498-396
 :   Mopec
 :  BD136
ZawBones Kit and Replacement Components, Mortech®
  76498-396
 :  Mopec
ZawBones Kit and Replacement Components, Mortech®
ZawBones Kit and Replacement Components, Mortech®
ZawBones fine replacement blade
$87.18
Each
All stainless steel construction.
 : ZawBones set includes (1) ZawBones stainless steel base with lower jaw, (2) ZawBones stainless steel upper jaws, (1) Stanley 15-113 contractor grade high tension hacksaw, (1) double hacksaw coarse tooth blade, (1) double hacksaw fine tooth blade, and (8) 3M SJ-5008 black bumpons.
  • Description:
    ZawBones fine replacement blade