Orders LinkContactUsLinkComponent
ZawBones Kit and Replacement Components, Mortech®
ZawBones Kit and Replacement Components, Mortech®
ZawBones coarse replacement blade
Catalog # 76498-394
Supplier:   Mopec
Supplier #:  BD135
ZawBones Kit and Replacement Components, Mortech®
Catalog # 76498-394
Supplier:  Mopec
ZawBones Kit and Replacement Components, Mortech®
ZawBones Kit and Replacement Components, Mortech®
ZawBones coarse replacement blade
Price
$87.18
Log in for availability
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All stainless steel construction.
Delivery information: ZawBones set includes (1) ZawBones stainless steel base with lower jaw, (2) ZawBones stainless steel upper jaws, (1) Stanley 15-113 contractor grade high tension hacksaw, (1) double hacksaw coarse tooth blade, (1) double hacksaw fine tooth blade, and (8) 3M SJ-5008 black bumpons.
Specifications
  • Description:
    ZawBones coarse replacement blade