CARTRIDGE 10IN 0.1UM PTFE BF/226 SILCN
Catalog # 76495-354
Supplier: PARKER HANNIFIN TE
CARTRIDGE 10IN 0.1UM PTFE BF/226 SILCN
Catalog # 76495-354
Supplier: PARKER HANNIFIN TE
Supplier Number: ZCTP1-010C-PS
Frequently Bought Together
VWR® Lab Markers
Supplier: VWR International
VWR® Disposable Serological Pipettes
Supplier: VWR International
VWR® Broken Glass Disposal Cartons
Supplier: VWR International
VWR® Powder-Free Nitrile Examination Gloves
Supplier: VWR International
Clorox® Concentrated Germicidal Bleach
Supplier: Janitorial Supplies
KIMTECH SCIENCE® KIMWIPES™ Delicate Task Wipers, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
VWR® High-Performance Centrifuge Tubes with Flat or Plug Caps, PP
Supplier: VWR International
VWR® Disposable Petri Dishes
Supplier: VWR International
PURPLE NITRILE™ and PURPLE NITRILE-XTRA™ Examination Gloves, Kimberly-Clark
Supplier: Ansell Healthcare
VWR® Nitrile Examination Gloves, Black
Supplier: VWR International
VWR® Universal Pipette Tips
Supplier: VWR International
VWR® Microcentrifuge Tubes, PP
Supplier: VWR International