Orders LinkContactUsLinkComponent
Test Tubes, Caplugs
5.0 ml, PP, Red, 12×75 mm, Round bottom
Catalog # 76493-680
Supplier:   Caplugs
 :  214-2367-R30
Test Tubes, Caplugs
Catalog # 76493-680
Supplier:  Caplugs
Test Tubes, Caplugs
Test Tubes, Caplugs
5.0 ml, PP, Red, 12×75 mm, Round bottom
Price
$77.74
Log in for availability
Carton of 1,000
Product Details & Documents
Test tubes provide clarity and can be used for mild acidic or alkali solutions, alcohol and salt, with exceptions of oxidizing acids and high concentrate aclohols.
  • Provided with or without colors
  • Tubes are bulk packed, non-sterile
  • Polystyrene tubes withstand up to 8200×g
  • Polypropylene tubes withstand up to 10000×g
  • 12×75 mm tubes are available with internally ribs for use in antibody coating and/or protein adsorptions, six slanted ribs increase useful surface area
The polypropylene tubes are chemically resistant and are appropriate for use with organic solvents and stronger acids, alkalis, alcohols, and chlorinated and aromatic hydrocarbons.
Specifications
  • Capacity:
    5.0 ml
  • Color:
    Red
  • Material:
    PP
  • Diameter ×H:
    12×75 mm
  • Bottom style:
    Round bottom