Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Test Tubes, Caplugs

Test Tubes, Caplugs

21 ml, PS, Natural, 16×150 mm, Green, PE screw cap, Round bottom
Catalog # 76493-664
Supplier:   Caplugs
Supplier #:  214-2099-G50
Test Tubes, Caplugs
Catalog # 76493-664
Supplier:  Caplugs
Test Tubes, Caplugs
Test Tubes, Caplugs

21 ml, PS, Natural, 16×150 mm, Green, PE screw cap, Round bottom

Price
$264.67
Log in for availability
Carton of 1,000
Product Details & Documents

Test tubes provide clarity and can be used for mild acidic or alkali solutions, alcohol and salt, with exceptions of oxidizing acids and high concentrate aclohols.

  • Provided with or without colors
  • Tubes are bulk packed, non-sterile
  • Polystyrene tubes withstand up to 8200×g
  • Polypropylene tubes withstand up to 10000×g
  • 12×75 mm tubes are available with internally ribs for use in antibody coating and/or protein adsorptions, six slanted ribs increase useful surface area

The polypropylene tubes are chemically resistant and are appropriate for use with organic solvents and stronger acids, alkalis, alcohols, and chlorinated and aromatic hydrocarbons.

Specifications
  • Cap type:
    PE screw cap
  • Capacity:
    21 ml
  • Color:
    Natural
  • Material:
    PS
  • Diameter ×H:
    16×150 mm
  • Cap color:
    Green
  • Bottom style:
    Round bottom