Test Tubes, Caplugs
16.0 ml, PS, Natural, 17×100 mm, Round bottom
Catalog # 76493-656
Supplier: Caplugs
Supplier #: 214-2070-010
Jump to:
Product Details & Documents
Test tubes provide clarity and can be used for mild acidic or alkali solutions, alcohol and salt, with exceptions of oxidizing acids and high concentrate aclohols.
- Provided with or without colors
- Tubes are bulk packed, non-sterile
- Polystyrene tubes withstand up to 8200×g
- Polypropylene tubes withstand up to 10000×g
- 12×75 mm tubes are available with internally ribs for use in antibody coating and/or protein adsorptions, six slanted ribs increase useful surface area
The polypropylene tubes are chemically resistant and are appropriate for use with organic solvents and stronger acids, alkalis, alcohols, and chlorinated and aromatic hydrocarbons.
Specifications
- Capacity:16.0 ml
- Color:Natural
- Material:PS
- Diameter ×H:17×100 mm
- Bottom style:Round bottom
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



