Order Entry
Puerto Rico
ContactUsLinkComponent
VACUM FILT ASY 47MM HOLD 2L FLASK 1L FUN
Catalog # 76491-132
Supplier:  Foxx Life Sciences
undefined
VACUM FILT ASY 47MM HOLD 2L FLASK 1L FUN
Catalog # 76491-132
Supplier:  Foxx Life Sciences
Supplier Number:  5350A30