Orders LinkContactUsLinkComponent
MXP Data Loggers, Cryopak Verification Technologies
MXP Data Loggers, Cryopak Verification Technologies
Data logger, MXP multi use
Catalog # 76490-422
Supplier #:  MXP-M
MXP Data Loggers, Cryopak Verification Technologies
Catalog # 76490-422
MXP Data Loggers, Cryopak Verification Technologies
MXP Data Loggers, Cryopak Verification Technologies
Data logger, MXP multi use
Price
$76.69
Log in for availability
Each
Product Details & Documents
The MXP data logger offers flexibility and longevity with a light sensor to detect and log if package is opened.
  • Long shelf life
  • Event logging feature
  • Internal digital temperature sensor
With its USB connectivity, the MXP data logger provides detailed reports in both PDF and Excel format, and is user friendly for both Windows and smartphone app use.
Specifications
  • Description:
    Data logger, MXP multi use