Objectives for REBEL Microscopes, ECHO
40× Plan Achromat Phase, LWD, 2 mm, N.A. 0.65
Catalog # 76490-286
Supplier: DISCOVER ECHO INC.
Supplier #: RMAG0433
Jump to:
Product Details & Documents
Specifications
- Description:40× Plan Achromat Phase, LWD
- Aperture:N.A. 0.65
- Working distance:2 mm