Objectives for REVOLVE Microscopes, Echo
Apochromat oil objective, 0.13 mm, 1.45 NA
Catalog # 76490-280
Supplier #: JMAG9001
Jump to:
Discontinued
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:100×
- Description:Apochromat oil objective
- Aperture:1.45 NA
- Working distance:0.13 mm