Objectives for REVOLVE Microscopes, Echo
Apochromat oil phase objective, 0.15 mm, 1.42 NA
Catalog # 76490-278
Supplier #: JMAG9602
Jump to:
Discontinued
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:60×
- Description:Apochromat oil phase objective
- Aperture:1.42 NA
- Working distance:0.15 mm