Orders LinkContactUsLinkComponent
Objectives for REVOLVE Microscopes, Echo
Objectives for REVOLVE Microscopes, Echo
Apochromat oil objective, 0.15 mm, 1.42 NA
Catalog # 76490-276
Supplier #:  JMAG9601
Objectives for REVOLVE Microscopes, Echo
Catalog # 76490-276
Supplier:  
Objectives for REVOLVE Microscopes, Echo
Apochromat oil objective, 0.15 mm, 1.42 NA
Discontinued
Product Details & Documents
Documents
Specifications
  • Magnification:
    60×
  • Description:
    Apochromat oil objective
  • Aperture:
    1.42 NA
  • Working distance:
    0.15 mm