Objectives for REVOLVE Microscopes, Echo
Apochromat objective, 0.6 mm, 0.80 NA
Catalog # 76490-270
Supplier #: JMAG9201
Jump to:
Discontinued
Product Details & Documents
Documents
Echo Revolve
Product Literature
Specifications
- Magnification:20×
- Description:Apochromat objective
- Aperture:0.80 NA
- Working distance:0.6 mm