TraceableLive® Subscription Coupon Codes
Product Details & Documents
Simplified cloud monitoring.
- TraceableLive®-capable products securely monitor critical environments and notify smartphones, tablets, or PCs if anything goes wrong
- Notifications for peace of mind
- All team members can receive email, text, and push notifications on the environments monitored
- Remotely view data logging history, effortlessly generate reports, and control alarm parameters
- Traceable products come calibrated to NIST standards to meet quality and regulatory requirements
- Digital data reports sent with ease whenever needed
- Customize reports with location, device, date range, and report format
- No limit to the number of devices on a TraceableLive® account for the Standard and Premium plans
Purchase a subscription code for one or two years of TraceableLive®. Subscriptions are available in standard or premium formats and are required per device. Scan the QR code to begin setup or go to traceablelive.com/activate and enter the unique 11 digit number on the card.
A standard subscription includes the following features: E-mail and Push Alarms, Unlimited Data Storage, Unlimited Team Members, Calibration Due notification, SMS (Text) - Not included with the subscription code, add on in the TraceableLive® app.
A premium subscription includes the following features: E-mail and Push Alarms, Free SMS (Text) Notifications, Unlimited Data Storage, Unlimited Team Members, Calibration Due notification, 21 CFR Part 11 Compliance.
Documents
Specifications
- Description:TraceableLive® 1 year premium subscription coupon code