Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
Flow Cell Priming Kit

Flow Cell Priming Kit

Flow cell priming kit, R9.4.1 flow cells, 6
Catalog # 76487-130
Flow Cell Priming Kit
Catalog # 76487-130
Supplier:  
CAS Number:  
Flow Cell Priming Kit
Flow Cell Priming Kit

Flow cell priming kit, R9.4.1 flow cells, 6

CAS Number:  
Discontinued
Product has been discontinued. However we found similar alternative products.
Alternatives
FLOW CELL PRIMING KIT V14 Catalog #76636-302

Product 76636-302 is similar to 76487-130 but manufacturer and specifications may vary.

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

Additional flow cell priming reagents to enable additional loading of more library during a sequencing run.

  • The kit can be used to prime a MinION™ Flow Cell for sequencing or to add more fuel to a flow cell during a sequencing experiment.

Flow Cell Priming Kit V14 (EXP-FLP004) ― the Flow Cell Priming Kit V14 contains Flow Cell Flush (FCF) and Flow Cell Tether (FCT), sufficient for 6 reactions. This kit is compatible with the latest library preparation kit chemistry (V14) and R10.4.1 Flow Cells.
Note: This kit is not compatible with the Ultra-Long DNA Sequencing Kit V14, which requires specific flow cell priming reagents.

Delivery information: Kits are shipped at 2 to 8 °C in an environmentally-friendly temperature-controlled shipping box. Upon receipt, please place the product in a long-term storage location at ‒20 °C. Oxford Nanopore Technologies deem the useful life of the product to be 3 months from receipt by the customer.

Caution: Oxford Nanopore Technologies products are not intended for use for health assessment or to diagnose, treat, mitigate, cure, or prevent any disease or condition.

Specifications

Specifications

  • Description:
    Flow cell priming kit
  • Compatible with:
    R9.4.1 flow cells
  • No. of reactions:
    6

Specifications