Basic Vortex Mixer, Thermo Scientific
Product 77041-354 is a replacement for 76485-986 and has the same packaging and specifications, but may be a different quantity.
Product Details & Documents
Thermo Scientific™ Basic Vortex Mixer is ideal for quick, uniform mixing of samples with precise speed control.
- Digital speed display
- Continuous and touch modes
- Speed range: 200 to 3000 rpm
- Speed accuracy: ±5% rpm
- Convenient speed adjustments by knob control
- Stable for long-term operations
- Multiple accessories for different vessels
Basic Vortex Mixer provides comprehensive mixing capabilities for enzymatic and RIA assays, general test tube mixing, biochemical assays, viral dilution preparation, precipitated assays, cell suspension vortexing and tissue sample mixing.
Motor type: DC motor; Orbit: 4 mm; Material: ABS housing
Certifications: cCSAus.
Ordering information: 2 years of warranty.
Specifications
- Capacity:Up to 19 tubes
- Description:Basic vortex mixer
- L×W×H:290×270×240 mm (11.4×10.7×9.4")
- Electrical:100/240 V, 50/60 Hz
- Shipping weight:6.8 kg (14.99 lbs.)
- Control type:Touch, continous
- Speed range:200 - 3000 rpm
- Max. load:0.5 kg
- Weight:6.5 kg
- Voltage:100 – 240 V
- Cat. No.:76485-986
- Orbit:4 mm
Accessories
Product Information
- Catalog #AvailabilityPrice
- Rubber bands, 6 per pack
Product Information
- Catalog #AvailabilityPrice
- Multi-power adaptor, 220 V, EU/UK/CHN plug
Product Information
- Catalog #AvailabilityPrice
- Power adaptor, 120 V, US plug
Product Information
- Catalog #For use withAvailabilityPrice
- Platform for flat bottomed vessels, 87.5×10 mmPlatform for flat bottomed vessels, 87.5×10 mm76417-608Digital vortex mixerLog in for availabilityEach - $33.59
Product Information
- Catalog #For use withAvailabilityPrice
- Tube holder, 10 mm Ø×19
Product Information
- Catalog #For use withAvailabilityPrice
- Microplate tray, black, 136.1×97.3×15 mmMicroplate tray, black, 136.1×97.3×15 mm76417-610Digital vortex mixerLog in for availabilityEach - $33.59
Product Information
- Catalog #For use withAvailabilityPrice
- Tube holder, 12 mm Ø×13
Product Information
- Catalog #For use withAvailabilityPrice
- Tube holder, 15 mm Ø×12
Product Information
- Catalog #For use withAvailabilityPrice
- Tube holder, 20 mm Ø×7
Product Information
- Catalog #For use withAvailabilityPrice
- Tube holder, 26mm Ø×4
Product Information
- Catalog #For use withAvailabilityPrice
- Rubber platform, black, 82×16.5 mmRubber platform, black, 82×16.5 mm76417-606Digital vortex mixerLog in for availabilityEach - $33.59
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



